Tisk…

3-Benzylidene-2,4-pentanedione 97%

Dodavatel: Thermo Fisher Scientific

L00623.06 L00623.14
L00623.06EA 1250 CZK
L00623.06 L00623.14
3-Benzylidene-2,4-pentanedione 97%
3-Benzylidene-2,4-pentanedione

Purity: 97% CAS No.: 4335-90-4 Molecular Formula: C12H12O2 Formula Weight: 188.23 Flash Point: 110°(230°F) Boiling Point: 185-188°/15mm Beilstein Reference: 972849 MDL No.: MFCD00014990

Vzorec: C₁₂H₁₂O₂
Bod varu: 185…188 °C (15 mmHg)
Hustota: 1,093
Bod vzplanutí: >110 °C (230 °F)
Storage Temperature: Ambient
MDL Number: MFCD00014990
CAS číslo: 4335-90-4
EINECS: 224-382-5

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR