Tisk…
Safety cabinets, poisons
Dodavatel: KOETTERMANN
379-00003 379-00001 379-00000 379-00002
KOTT379-00003EA 10820 CZK
KOTT379-00003 KOTT379-00001 KOTT379-00000 KOTT379-00002
Safety cabinets, poisons
Nábytek kancelářský Skříně pro zásoby
These cabinets are available with normal and hinged doors.
  • Lockable
  • Continuous extraction inside the cabinet
  • Can be wall mounted or installed in chemical storage cabinet
Epoxy powder coated steel.