Tisk…
Weighing pans, aluminium
Dodavatel: KOEHLER TECHNISCHE PRODUKTEN
2-1607
KOEH2-1607EA 1420 CZK
KOEH2-1607
Weighing pans, aluminium
Lodičky Lodičky navažovací
Made of Aluminium, bright.
  • Good temperature resistance
  • Good chemical resistance
  • Lightweight
  • Stackable