Tisk…
Equipotential bonding electrode
Dodavatel: KNICK
ZU0073
KNICZU0073EA 15260 CZK
KNICZU0073
Equipotential bonding electrode
Elektrody Elektrody ostatní
Electrode for monitoring the reference system of the pH sensor or for simultaneous ORP measurements.