Tisk…
Embedding cassettes, without lid
Dodavatel: KLINIPATH
310-04T
KLIN310-04TEA 9380 CZK
KLIN310-04T
Embedding cassettes, without lid
Kazety pro zalévání vzorků Kazety tkáňové
Every cassette maintain its stability, rigidness and shape during processing of tissues.
Balení: Hoppers contain 75 standard cassettes and are specially packed for Thermo Fisher® cassette printers. Taped stacks contain 40 cassettes and are specially packed for Sakura® / Leica® cassette printers.