Tisk…
Embedding cassettes with disposable lid
Dodavatel: KLINIPATH
110-02T 110-06T 110-02S
KLIN110-02TEA 4820 CZK
KLIN110-02T KLIN110-06T KLIN110-02S
Embedding cassettes with disposable lid
Kazety pro zalévání vzorků Kazety tkáňové
Embedded cassettes with hinged lid.
  • Cassettes maintain their stability, rigidness and shape during processing of tissues
Informace pro objednávání: Available in different colours.
Balení: Hoppers contain 80 cassettes which are specially packed for Thermo Fisher® cassette printers. Taped stacks contain 40 cassettes which are specially packed for Sakura® / Leica® cassette printers. Lids are packed separately.