Tisk…
Wipe rolls, WypAll® L10 compact food and hygiene
Dodavatel: KIMBERLY CLARK
7225 7236
KIMB7225EA 6700 CZK
KIMB7225 KIMB7236
Wipe rolls, WypAll® L10 compact food and hygiene
Ubousky/Utěrky Utěrky jednorázové a ručníky Ubrousky pro běžné aplikace
Compact disposable paper wipes are 1-ply and designed for light-duty wiping in areas such as food preparation and service surfaces.
  • Compact roll design is small, portable, and easy to handle
  • Ease of use helps to support hygiene standards and compliance while also saving space
  • WypAll® wiping paper is designed to be used once with detergent or disinfectant
These dry cleaning wipes have been tested as compatible for use in spray and wipe application with peracetic acid, hydrogen peroxide, and quats (quaternary ammonium compounds). Spray and wipingMore Product Information