Tisk…
Cleaning cloths, undyed, WYPALL*
Dodavatel: KIMBERLY CLARK
38715
KIMB38715EA 4780 CZK
KIMB38715
Cleaning cloths, undyed, WYPALL*
Ubousky/Utěrky Utěrky čistící pro opakovatelné použití Utěrky pro běžné aplikace
These microfibre cloths lift and trap surface dirt. Ideal for cleaning in offices, healthcare, and hospitality.
  • Perfect for use in transportation manufacturing
  • Suitable for a wide range of cleaning tasks
  • Washable
These microfibre cloths are designed with a high quality polyester/polyamide that doesn't fray making them perfect for automotive valet tasks.