Tisk…
Cold trap, type S
Dodavatel: KGW
17407 17405
KGWA17407EA 12830 CZK
KGWA17407 KGWA17405
Cold trap, type S
Pasti vakuové
Type S cold trap has a conical ground joint NS 45/40 as connection part. The condensation area can be separated from the upper section of cold trap. Therefore it is easy to pour out the condensate and to clean the whole cold trap. The connection part NS 45/40 is secured by a spring clamp out of steel.
Certifikace: Made from borosilicate glass 3.3, in compliance with DIN/ISO 3585.