Tisk…
Balance boats, disposable
Dodavatel: KARTELL
11105 11103 11104 11105 1110003 1110103 1110109 1110203 1110209
611-1393EA 3000 CZK
611-1393 KART11103 KART11104 KART11105 KART1110003 KART1110103 KART1110109 KART1110203 KART1110209
Balance boats, disposable
Lodičky Lodičky navažovací
Balance boats, high impact PS.
  • Easily fashioned to make a funnel shape
  • The DS model is available in either white or black so the operator can easily see that no trace of sample remains adhering to the boat
  • Reproducible tare weight: batch to batch variation is normally less than 5%
  • Antistatic
Diamond (DS) and square shaped (SQ) lightweight weighing boats provide an easy and inexpensive system for weighing any medium with non contaminating properties (made of HIPS with hydrophobic surface).