Tisk…
Sterilisation Wraps, 2-Ply SMS, One-Step Super Duty
Dodavatel: KEYSTONE CLEANROOM PRODUCTS
KCRP1-600-0808 KCRP1-600-0909 KCRP1-600-0912 KCRP1-600-1212 KCRP1-600-1515 KCRP1-600-1818 KCRP1-600-1822 KCRP1-600-2020 KCRP1-600-2424 KCRP1-600-3030 KCRP1-600-3636 KCRP1-600-4040 KCRP1-600-4545 KCRP1-600-4548 KCRP1-600-4848 KCRP1-600-5454 KCRP1-600-5472 KCRP1-600-5490 KCRP1-600-6060 KCRP1-600-6080
129-3813EA 14680 CZK
129-3813 129-3814 129-3815 129-3816 129-3817 129-3818 129-3819 129-3820 129-3992 129-3993 129-3994 129-3995 KACCKCRP1-600-4545 129-3996 129-3997 129-3998 129-3999 129-4004 129-4005 129-4006
Sterilisation Wraps, 2-Ply SMS, One-Step Super Duty
Utěrky sterilizační
One-Step heavy duty SMS 2-ply wrappers provide an excellent microbial barrier. Ideal for pharmaceutical production applications.
  • Manufactured and packaged in a certified ISO 8 Cleanroom
  • 2-ply
  • Clean - no cellulose
  • Strong, rip and tear resistant 50 gsm SMS material
  • An extensive range of sizes available means less cutting
  • Hydrophobic barrier allows sterilization gases, but prevents liquid and microbial penetration
  • Gram weight: 88 (each layer)
For use in steam, ethylene oxide and hydrogen peroxide sterilisation processes.
Informace o dodávce: More Product Information