Tisk…
Slide Closure Zip Lock Bags, Sterile
Dodavatel: KEYSTONE CLEANROOM PRODUCTS
1215SZLDPST2
KACC1215SZLDPST2EA 42690 CZK
KACC1215SZLDPST2
Slide Closure Zip Lock Bags, Sterile
Sáčky Obaly sterilizační
Pre-sterilised and packaged in multiple layers of Tyvek peel pouches and poly bags to gain access into the aseptic area without excessive spraying and wiping with sanitisation agents. The bags are used for the removal and transport of media plates from the aseptic area to QC laboratories.
  • Manufactured and packaged in an ISO certified cleanroom
  • Tear and puncture resistant 2,7 mill low density polyethylene
  • Each bag has an integral zip seal with the slide lock closure system
  • Two zip-lock bags are double packaged in Tyvek/film chevron peel pouches and then
More Product Information