Tisk…
Autoclavable Tyvek Breather Bags, Easy Chevron Peel and Self Seal Closure, Steam Indicator
Dodavatel: KEYSTONE CLEANROOM PRODUCTS
00308TWBBSSSD 01214TWBBSSSD 01217TWBBSSSD 00510TWBBSSSD 00612TWBBSSSD
KACC00308TWBBSSSDEA 18970 CZK
KACC00308TWBBSSSD KACC01214TWBBSSSD KACC01217TWBBSSSD KACC00510TWBBSSSD KACC00612TWBBSSSD
Autoclavable Tyvek Breather Bags, Easy Chevron Peel and Self Seal Closure, Steam Indicator
Sáčky Obaly sterilizační
A significant upgrade over paper-based steam sterilisation pouches. These Tyvek® peel pouches will not rip or puncture like paper-based autoclave pouches routinely do.
  • Manufactured and packaged in an ISO certified cleanroom Chevron peel opening Autoclavable at 121 to 125 °C Tear and puncture resistant Hydrophobic Lint-free Covenient self-sealing closure
The medical Grade 1073B Tyvek® material has the highest microbial barrier properties of any steam sterilization compatible materials. These pouches are also compatible with VHP Decontamination cycles
More Product Information