Tisk…
Clear safety spectacles, Crackerjack™
Dodavatel: JSP
1CRAK23C
JSPA1CRAK23CEA 172 CZK
JSPA1CRAK23C
Clear safety spectacles, Crackerjack™
Brýle
Stylish, single lens, lightweight safety spectacle.
  • Ultra enveloping
  • Lateral protection
  • Flat temples
The Crackerjack™ is a stylish single lens spectacle that is lightweight with flat temples providing comfort for extended periods of use. Its design makes it a perfect fit for wearing in conjunction with helmets and ear defenders.
Certifikace: Meets EN 166:2001.T9:V9