Tisk…
Tees and crosses, stainless steel
Dodavatel: VICI JOUR RESEARCH
JR-ZT1 JR-ZX1M JR-ZX1C JR-ZX1 JR-ZT1M
JOURJR-ZT1EA 0 CZK
JOURJR-ZT1 JOURJR-ZX1M JOURJR-ZX1C JOURJR-ZX1 JOURJR-ZT1M
Tees and crosses, stainless steel
Fitinky a konektory
These stainless steel 316 tees and crosses are ideal for joining solvent lines.
  • Choice of three bores
  • Low dead volume
  • Pressure rating: <1375 bar (20,000 psi)
They are precision machined with a focus on the concentricity of the bore as well as on the finish of the surfaces to gave an absolute leak-free connection. Internal threads make stronger connections and internal fitting details provide the lowest dead volume for high performance instrumentation.
Informace o dodávce: Four nuts and ferrules with each tee.