Tisk…
Safety air outlet filter
Dodavatel: VICI JOUR RESEARCH
JR-S-20004
JOURJR-S-20004EA 1430 CZK
JOURJR-S-20004
Safety air outlet filter
Filtry Filtry pro práci s kapalinami
This safety air outlet filter regulates the pressure during helium sparging.