Tisk…

Crocein scarlet 7B ≥70% (dye content)

Dodavatel: Thermo Fisher Scientific

J66876.03 J66876.06 J66876.14
J66876.03EA 858 CZK
J66876.03 J66876.06 J66876.14
Crocein scarlet 7B ≥70% (dye content)
Crocein scarlet 7B

CAS No.: 6226-76-2 Molecular Formula: C24H18N4Na2O7S2 Formula Weight: 584.53 Physical Form: Powder Appearance: Red Melting Point: 181-188° Solubility: Soluble in water MDL No.: MFCD00056588

Vzorec: C₂₄H₁₈N₄Na₂O₇S₂
Bod tání: 181…188 °C
Storage Temperature: Ambient
MDL Number: MFCD00056588
CAS číslo: 6226-76-2

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR