Tisk…

Gatifloxacin sesquihydrate

Dodavatel: Thermo Fisher Scientific
Varování

Synonyms: 1-Cyclopropyl-6-fluoro-8-methoxy-7-(3-methylpiperazin-1-yl)-4-oxo-1,4-dihydroquinoline-3-carboxylic acid sesquihydrate

J66036.03 J66036.06
J66036.03EA 1330 CZK
J66036.03 J66036.06
Gatifloxacin sesquihydrate
Gatifloxacin sesquihydrate

CAS No.: 180200-66-2 Molecular Formula: C19H22FN3O4.1.5H2O Formula Weight: 402.42 Physical Form: Powder Appearance: White to pale yellow Boiling Point: 684° Solubility: Soluble in 1N NaOH at 10mg/ml MDL No.: MFCD22373645


Applications: A broad spectrum antibiotic

Vzorec: C₃₈H₅₀F₂N₆O₁₁
MW: 804.83 g/mol
Bod varu: 684 °C
Storage Temperature: Ambient
MDL Number: MFCD22373645
CAS číslo: 180200-66-2

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR