Tisk…

IPA-3

Dodavatel: Thermo Fisher Scientific
Nebezpečí

Synonyms: p21-Activated Kinase Inhibitor III, 1,1'-Disulphanediylbis(naphthalen-2-ol), 1,​1'-​dithiobis-​2-​naphthalenol

J65848.MAEA 6150 CZK
J65848.MA
IPA-3
IPA-3

p21-Activated Kinase Inhibitor III, also known as IPA-3, is an allosteric inhibitor of p21 activated kinase (Pak1). It binds to the autoinhibitory domain of Pak1, and has been implicated in tumorigenesis and metastasis.


  • Allosteric inhibitor of p21 (PAK1)
  • Implicated in tumorigenesis and metastasis
  • Soluble in DMSO or ethanol.
  • Insoluble in water


Yellow


Soluble in DMSO or ethanol. Insoluble in water.

Vzorec: C₂₀H₁₄O₂S₂
MW: 350.45 g/mol
Storage Temperature: Freezer
MDL Number: MFCD00388917
CAS číslo: 42521-82-4
UN: 3077
ADR: 9 III

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR