Tisk…

3'-Deoxyuridine

Dodavatel: Thermo Fisher Scientific

J65531.MB
J65531.MBEA 12630 CZK
J65531.MB
3'-Deoxyuridine
3'-Deoxyuridine

CAS No.: 7057-27-4 Molecular Formula: C9H12N2O5 Formula Weight: 228.21 Physical Form: Powder Appearance: White to off-white Solubility: Soluble in DMSO, methanol, or water. Also soluble in dichloromethane MDL No.: MFCD00079153


Applications: A potential anti-cancer and anti-viral agent

Vzorec: C₉H₁₂N₂O₅
MW: 228.21 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00079153
CAS číslo: 7057-27-4

Objednat

Produktová specifikace

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR