Tisk…

8-Amino-2-naphthalenesulphonic acid 95%

Dodavatel: Thermo Fisher Scientific
Nebezpečí

Synonyms: 1-Naphthylamine-7-sulphonic acid, 1,7-Cleve's acid

J65304.14 J65304.22
J65304.14EA 2370 CZK
J65304.14 J65304.22
8-Amino-2-naphthalenesulphonic acid 95%
8-Amino-2-naphthalenesulphonic acid
Vzorec: C₁₀H₉NO₃S
MW: 223.25
Bod tání: >300 °C
Storage Temperature: Ambient
MDL Number: MFCD00004032
CAS číslo: 119-28-8
EINECS: 204-311-4
UN: 2585
ADR: 8 III
Merck Index: 142,351

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR