Tisk…

3,3',5,5'-Tetramethylbenzidin, Liquid, Biotechnology Grade component_1

Dodavatel: VWR Chemicals

Synonyms: TMB, 3,3',5,5'-Tetramethyl-[1,1'-biphenyl]-4,4'-diamine

J644-100MLEA 3790 CZK
J644-100ML
3,3',5,5'-Tetramethylbenzidin, Liquid, Biotechnology Grade component_1
3,3',5,5'-Tetramethylbenzidin

Ready-to-use sensitive substrate for the detection of horseradish peroxidase activity


Absorbs at 450 nm (yellow end-product). Ideal for ELISA and solution assays.

Vzorec: C₁₆H₂₀N₂
MW: 240.35
Bod varu: 402
MDL Number: MFCD00007748
CAS číslo: 54827-17-7

Objednat

Výsledky specifikačního testu

Abs.@650nm <= 0.02
pH @25°C 5.0 - 7.0
UV/Vis Assay PASS / Fail


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR