Tisk…

Clotrimazole

Dodavatel: Thermo Fisher Scientific
Varování

J63895.06 J63895.14
J63895.06EA 2820 CZK
J63895.06 J63895.14
Clotrimazole
Clotrimazole

CAS No.: 23593-75-1 Molecular Formula: C22H17ClN2 Formula Weight: 344.84 Physical Form: Powder Appearance: White to off-white Melting Point: 141-149° Solubility: Soluble in chloroform, DMSO, DMF and alcohols MDL No.: MFCD00057220


Applications: Specific inhibitor of calcium-activated potassium channels

Vzorec: C₂₂H₁₇ClN₂
Bod tání: 141…149 °C
Storage Temperature: Ambient
MDL Number: MFCD00057220
CAS číslo: 23593-75-1
EINECS: 245-764-8
Merck Index: 14,2417

Objednat

Produktová specifikace

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR