Tisk…

Buffer, TBE solution 10X concentrate (TRIS-borate-EDTA buffer)

Dodavatel: Thermo Fisher Scientific

J62449.K2 J62449.K3 J62449.K7
J62449.K2EA 1710 CZK
J62449.K2 J62449.K3 J62449.K7
Buffer, TBE solution 10X concentrate (TRIS-borate-EDTA buffer)
Pufry Pufry pro elektroforézu Pufry do dělícího gelu

Physical Form: Liquid
Appearance: Colourless
MDL No.: MFCD00236358

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR