Tisk…

L(+)-Serin cell culture reagent

Dodavatel: Thermo Fisher Scientific

Synonyms: H-Ser-OH, (S)-2-Amino-3-hydroxypropanoic acid, Serine, (S)-(+)-Serine

J62187.09 J62187.22 J62187.36 J62187.A1
J62187.09EA 1220 CZK
J62187.09 J62187.22 J62187.36 J62187.A1
L(+)-Serin cell culture reagent
L(+)-Serin

CAS No.: 56-45-1 Molecular Formula: C3H7NO3 Formula Weight: 105.09 Physical Form: Powder Appearance: White Melting Point: 222° Beilstein Reference: 1721404 Merck Reference: 14,8460 MDL No.: MFCD00064224

Vzorec: C₃H₇NO₃
MW: 105.09
Bod varu: 259
Bod tání: 222 °C
Hustota: 1.6 g/cm³ (22 °C)
Storage Temperature: Ambient
MDL Number: MFCD00064224
CAS číslo: 56-45-1
EINECS: 200-274-3
Merck Index: 14,8460

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR