Tisk…

Adenosine-3',5'-cyclic monophosphate sodium salt 99%

Dodavatel: Thermo Fisher Scientific

J62174.MD J62174.03 J62174.06
J62174.MDEA 3780 CZK
J62174.MD J62174.03 J62174.06
Adenosine-3',5'-cyclic monophosphate sodium salt 99%
Adenosine-3',5'-cyclic monophosphate sodium salt

Purity: 99% CAS No.: 37839-81-9 Molecular Formula: C10H11N5NaO6P Formula Weight: 351.19 Storage Temperature: -30°C to -10°C Physical Form: Crystalline powder Appearance: White Beilstein Reference: 54612 MDL No.: MFCD00069736

Vzorec: C₁₀H₁₁N₅NaO₆P
Storage Temperature: Freezer
MDL Number: MFCD00069736
CAS číslo: 37839-81-9

Objednat

Produktová specifikace

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR