Tisk…

L(+)-Lyzin monohydrochlorid cell culture reagent

Dodavatel: Thermo Fisher Scientific

J62099.22 J62099.36 J62099.A1
J62099.22EA 1410 CZK
J62099.22 J62099.36 J62099.A1
L(+)-Lyzin monohydrochlorid cell culture reagent
L(+)-Lyzin monohydrochlorid

CAS No.: 657-27-2 Molecular Formula: C6H14N2O2.HCl Formula Weight: 182.65 Physical Form: Powder Appearance: White to off-white Beilstein Reference: 3563889 Merck Reference: 14,5636 MDL No.: MFCD00064564

Vzorec: H₂N(CH₂)₄CH(NH₂)CO₂H·HCl
Bod tání: 263…264 °C
Storage Temperature: Ambient
MDL Number: MFCD00064564
CAS číslo: 657-27-2
EINECS: 211-519-9
Merck Index: 14,5636

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR