Tisk…

Paraformaldehyd fixative solution

Dodavatel: Thermo Fisher Scientific
Varování

J61984.AK J61984.AP
J61984.AKEA 2900 CZK
J61984.AK J61984.AP
Paraformaldehyd fixative solution
Paraformaldehyd

Paraformaldehyde fixative solution finds application in proteins, peptides and the enzymes of low molecular weight.


Storage Temperature: 2°C to 8°C Physical Form: Liquid MDL No.: MFCD00133991

Vzorec: (CH₂O)n
Storage Temperature: Refrigerator
MDL Number: MFCD00133991
CAS číslo: 30525-89-4

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR