Tisk…

N-Succinimidyl 4-Maleimidobutyrate

Dodavatel: Thermo Fisher Scientific
Varování

Synonyms: GMBS

J61850.MCEA 8000 CZK
J61850.MC
N-Succinimidyl 4-Maleimidobutyrate
GMBS (N-Succinimidyl 4-maleimidobutyrate)

CAS No.: 80307-12-6 Molecular Formula: C12H12N2O6 Formula Weight: 280.23 Storage Temperature: -30°C to -10°C Physical Form: Powder Appearance: White Melting Point: 130-132° Beilstein Reference: 6427689 MDL No.: MFCD00036817


Applications: A heterobifunctional cross-linking reagent with amine and sulfhydryl reactivity. Useful in enzyme immunoassays

Vzorec: C₁₂H₁₂N₂O₆
MW: 280.24 g/mol
Bod tání: 130…132 °C
Storage Temperature: Freezer
MDL Number: MFCD00036817
CAS číslo: 80307-12-6

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR