Tisk…

Buffers, PIPES, 0,5 M solutions

Dodavatel: Thermo Fisher Scientific

J61786.AE J61786.AK J61224.AE J61224.AK J61406.AE J61406.AK J63234.AE J63234.AK J63617.AE J63617.AK
J61786.AEEA 2790 CZK
J61786.AE J61786.AK J61224.AE J61224.AK J61406.AE J61406.AK J63234.AE J63234.AK J63617.AE J63617.AK
Buffers, PIPES, 0,5 M solutions
Pufry Pufry biologické

Liquid


CAS No.: 5625-37-6
Appearance: Colorless
MDL No.: MFCD00006159


Upozornení: Storage Temperature: 2 to 8 °C

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR