Tisk…

Tris-(hydroxymethyl)aminomethan 99.9% for molecular biology

Dodavatel: Thermo Fisher Scientific
Varování

Synonyms: 2-Amino-2-(hydroxymethyl)propane-1,3-diol(tris), Tromethamine, Tris(hydroxymethyl)methylamine, 2-Amino-2-(hydroxymethyl)propane-1,3-diol, 2-Amino-2-(hydroxymethyl)-1,3-propanediol, Trizma(R) base, TRIS base, THAM

J61062.22 J61062.36
J61062.22EA 1970 CZK
J61062.22 J61062.36
Tris-(hydroxymethyl)aminomethan 99.9% for molecular biology
Tris-(hydroxymethyl)aminomethan
Vzorec: NH₂C(CH₂OH)₃
Bod varu: 219…220 °C (10 mmHg)
Bod tání: 168…172 °C
Hustota: 1,353
Storage Temperature: Ambient
MDL Number: MFCD00004679
CAS číslo: 77-86-1
EINECS: 201-064-4
Merck Index: 14,9772
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=11a03d1ab6b2c11b9a2c83f0a73fc4264cec0110&vid=5ce890cdf298a59a7900a07fee1c857a5e5e61e0&prid=web&clid=SAPDAM

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR