Tisk…

Octyl-β-D-thioglucopyranoside ≥98%

Dodavatel: Thermo Fisher Scientific

J61028.06 J61028.09 J61028.88
J61028.06EA 10130 CZK
J61028.06 J61028.09 J61028.88
Octyl-β-D-thioglucopyranoside ≥98%
Octyl-β-D-thioglucopyranoside

Purity: 98+% CAS No.: 85618-21-9 Molecular Formula: C14H28O5S Formula Weight: 308.44 Physical Form: Crystalline powder Appearance: White to off-white Melting Point: 128° Solubility: Soluble in ethanol, water or methanol Beilstein Reference: 4182783 MDL No.: MFCD00012189


Non-ionic detergent used for the solubilization of membrane proteins

Vzorec: C₁₄H₂₈O₅S
Bod tání: 128 °C
Storage Temperature: Ambient
MDL Number: MFCD00012189
CAS číslo: 85618-21-9

Objednat

Produktová specifikace

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR