Tisk…

Buffers, potassium phosphate, 0,5 M solutions

Dodavatel: Thermo Fisher Scientific

J61553.AK J61553.AP J60326.AK J60326.AP J60584.AK J60584.AP J61413.AK J61413.AP
J61553.AKEA 2610 CZK
J61553.AK J61553.AP J60326.AK J60326.AP J60584.AK J60584.AP J61413.AK J61413.AP
Buffers, potassium phosphate, 0,5 M solutions
Pufry Pufry biologické

CAS No.: 7778-77-0
Physical Form: Liquid
MDL No.: MFCD00011401

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR