Tisk…

β-Nicotinamide adenine dinucleotide phosphate reduced tetrasodium salt hydrate 97%

Dodavatel: Thermo Fisher Scientific

Synonyms: β-NADPH tetrasodium salt tetrahydrate

J60387.MCEA 7380 CZK
J60387.MC J60387.ME
β-Nicotinamide adenine dinucleotide phosphate reduced tetrasodium salt hydrate 97%
β-Nicotinamide adenine dinucleotide phosphate reduced tetrasodium salt hydrate

Applications: beta-Nicotinamide adenine dinucleotide phosphate reduced tetrasodium salt hydrate is used as NADP+/NADPH redox pair is used in a variety of antioxidation mechanism where it protects agains reactive oxidation species accumulation. NADPH is generated in vivio by the pentose phosphate pathway.


  • Store in cool place
  • Keep container tightly closed in a dry and well-ventilated place
  • Light sensitive
  • Hygroscopic


Purity: 98+%
Molecular Formula: C21H26N7Na4O17P3.xH2O
Formula Weight: 905.42 (833.35anhy)
Storage Temperature: -30°C to -10°C
Physical Form: Powder
Appearance: White to off-white
MDL No.: MFCD07366294

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR