Tisk…
XELA UHPLC Columns, Silica
Dodavatel: ISERA
A211-411A00 A211-411B00 A211-411D00 A211-411F00 A211-411G00 A211-411H00 A211-411K00 A211-411M00 A211-411U00 A211-411V00 A211-411X00 A211-411Z00 A212-411A30
ISERA211-411A00EA 12590 CZK
ISERA211-411A00 ISERA211-411B00 ISERA211-411D00 ISERA211-411F00 ISERA211-411G00 ISERA211-411H00 ISERA211-411K00 ISERA211-411M00 ISERA211-411U00 ISERA211-411V00 ISERA211-411X00 ISERA211-411Z00 ISERA212-411A30
XELA UHPLC Columns, Silica
Kolony chromatografické
A maximum of separation efficiency and perfect selectivity comply with highest demands.
  • Optimal resolution is achieved by high separation efficiency and appropriate selectivity
  • Powerful
  • Fast and sensitive
  • UHPLC without limitation
XELA columns for UHPLC combine these two crucial parameters of efficency and selectivity in an excellent manner. Therefore, they are the perfect tool for successful LC separations in the high-performance section.
1,7 µm silica particles with a narrow particle size distribution and a homogeneous pore structure ensureMore Product Information
Objednat
Produktová specifikace
Popis Analytical column Guard column
Phase name XELA XELA
Environmentally Preferable
Maximální tlak 1000 bar 1000 bar
Rozsah pH 2,5 - 7 2,5 - 7
Kód USP L3 L3
Uskladnění RT RT
Podmínky k odeslání Shipped under testing eluent.