Tisk…
ISAspher HPLC Columns, C8 Select
Dodavatel: ISERA
A111-R25A00 A111-R25B00 A111-R25C00 A111-R25D00 A111-R25E00 A111-R25F00 A111-R25G00 A111-R25H00 A111-R25J00 A111-R25K00 A111-R25L00 A111-R25M00 A111-R25N00 A111-R25P00 A111-R25Q00 A111-R25R00 A111-R25S00 A111-R25T00 A111-R25U00 A111-R25V00 A111-R25W00 A111-R25X00 A111-R25Y00 A111-R25Z00
ISERA111-R25A00EA 6870 CZK
ISERA111-R25A00 ISERA111-R25B00 ISERA111-R25C00 ISERA111-R25D00 ISERA111-R25E00 ISERA111-R25F00 ISERA111-R25G00 ISERA111-R25H00 ISERA111-R25J00 ISERA111-R25K00 ISERA111-R25L00 ISERA111-R25M00 ISERA111-R25N00 ISERA111-R25P00 ISERA111-R25Q00 ISERA111-R25R00 ISERA111-R25S00 ISERA111-R25T00 ISERA111-R25U00 ISERA111-R25V00 ISERA111-R25W00 ISERA111-R25X00 ISERA111-R25Y00 ISERA111-R25Z00
ISAspher HPLC Columns, C8 Select
Kolony chromatografické
HPLC column of the ISAspher line guarantee the highest possible reliability in a wide range of applications.
  • The production of the silica used as well as the functionalisation are carried out according to highly developed and validated process
  • Reliable and proven
  • Flexibility and robustness for every HPLC application
This ensure unchanged properties of the stationary phases in terms of selectivity and consistent chemical properties.
Direct scale-up e.g. starting from a particle size of 5 µm to semi-preparative or preparative applications (up to 45 µMore Product Information