Tisk…
ISAspher HPLC Columns, PFP
Dodavatel: ISERA
A111-M25A00 A111-M25B00 A111-M25C00 A111-M25D00 A111-M25E00 A111-M25F00 A111-M25G00 A111-M25H00 A111-M25J00 A111-M25K00 A111-M25L00 A111-M25M00 A111-M25N00 A111-M25P00 A111-M25Q00 A111-M25R00 A111-M25S00 A111-M25T00 A111-M25U00 A111-M25V00 A111-M25W00 A111-M25X00 A111-M25Y00 A111-M25Z00 A112-M25A30 A112-M25B30 A112-M25C30 A112-M25E30 A112-M25F30 A112-M25G30
ISERA111-M25A00EA 7590 CZK
ISERA111-M25A00 ISERA111-M25B00 ISERA111-M25C00 ISERA111-M25D00 ISERA111-M25E00 ISERA111-M25F00 ISERA111-M25G00 ISERA111-M25H00 ISERA111-M25J00 ISERA111-M25K00 ISERA111-M25L00 ISERA111-M25M00 ISERA111-M25N00 ISERA111-M25P00 ISERA111-M25Q00 ISERA111-M25R00 ISERA111-M25S00 ISERA111-M25T00 ISERA111-M25U00 ISERA111-M25V00 ISERA111-M25W00 ISERA111-M25X00 ISERA111-M25Y00 ISERA111-M25Z00 ISERA112-M25A30 ISERA112-M25B30 ISERA112-M25C30 ISERA112-M25E30 ISERA112-M25F30 ISERA112-M25G30
ISAspher HPLC Columns, PFP
Kolony chromatografické
HPLC column of the ISAspher line guarantee the highest possible reliability in a wide range of applications.
  • The production of the silica used as well as the functionalisation are carried out according to highly developed and validated process
  • Reliable and proven
  • Flexibility and robustness for every HPLC application
This ensure unchanged properties of the stationary phases in terms of selectivity and consistent chemical properties.
Direct scale-up e.g. starting from a particle size of 5 µm to semi-preparative or preparative applications (up to 45 µMore Product Information