Tisk…
ISAspher HPLC Columns, Phenyl
Dodavatel: ISERA
A111-F23A00 A111-F23B00 A111-F23C00 A111-F23D00 A111-F23E00 A111-F23F00 A111-F23G00 A111-F23H00 A111-F23J00 A111-F23K00 A111-F23L00 A111-F23M00 A111-F23N00 A111-F23P00 A111-F23Q00 A111-F23R00 A111-F23S00 A111-F23T00 A111-F23U00 A111-F23V00 A111-F23W00 A111-F23X00 A111-F23Y00 A111-F23Z00 A111-F25A00 A111-F25B00 A111-F25C00 A111-F25D00 A111-F25E00 A111-F25F00 A111-F25G00 A111-F25H00 A111-F25J00 A111-F25K00 A111-F25L00 A111-F25M00 A111-F25N00 A111-F25P00 A111-F25Q00 A111-F25R00 A111-F25S00 A111-F25T00 A111-F25U00 A111-F25V00 A111-F25W00 A111-F25X00 A111-F25Y00 A111-F25Z00 A111-F29A00 A111-F29B00 A111-F29C00 A111-F29D00 A111-F29E00 A111-F29F00 A111-F29G00 A111-F29H00 A111-F29J00 A111-F29K00 A111-F29L00 A111-F29M00 A111-F29N00 A111-F29P00 A111-F29Q00 A111-F29R00 A111-F29S00 A111-F29T00 A111-F29U00 A111-F29V00 A111-F29W00 A111-F29X00 A111-F29Y00 A111-F29Z00 A113-F25A00 A113-F25B00 A113-F25D00 A113-F25H00 A113-F25J00 A113-F25K00 A113-F25L00 A113-F29A00 A113-F29B00 A113-F29D00 A113-F29H00 A113-F29J00 A113-F29K00 A113-F29L00 A112-F25A30 A112-F25B30 A112-F25C30 A112-F25E30 A112-F25F30 A112-F25G30 A112-F25A00 A112-F25B00 A112-F25C00 A112-F25E00 A112-F25F00 A112-F25G00 A112-F23A30 A112-F23B30 A112-F23C30 A112-F23E30 A112-F23F30 A112-F23G30 A112-F23A00 A112-F23B00 A112-F23C00 A112-F23E00 A112-F23F00 A112-F23G00 A112-F29A30 A112-F29B30 A112-F29C30 A112-F29E30 A112-F29F30 A112-F29G30 A112-F29A00 A112-F29B00 A112-F29C00 A112-F29E00 A112-F29F00 A112-F29G00
ISERA111-F23A00EA 8070 CZK
ISERA111-F23A00 ISERA111-F23B00 ISERA111-F23C00 ISERA111-F23D00 ISERA111-F23E00 ISERA111-F23F00 ISERA111-F23G00 ISERA111-F23H00 ISERA111-F23J00 ISERA111-F23K00 ISERA111-F23L00 ISERA111-F23M00 ISERA111-F23N00 ISERA111-F23P00 ISERA111-F23Q00 ISERA111-F23R00 ISERA111-F23S00 ISERA111-F23T00 ISERA111-F23U00 ISERA111-F23V00 ISERA111-F23W00 ISERA111-F23X00 ISERA111-F23Y00 ISERA111-F23Z00 ISERA111-F25A00 ISERA111-F25B00 ISERA111-F25C00 ISERA111-F25D00 ISERA111-F25E00 ISERA111-F25F00 ISERA111-F25G00 ISERA111-F25H00 ISERA111-F25J00 ISERA111-F25K00 ISERA111-F25L00 ISERA111-F25M00 ISERA111-F25N00 ISERA111-F25P00 ISERA111-F25Q00 ISERA111-F25R00 ISERA111-F25S00 ISERA111-F25T00 ISERA111-F25U00 ISERA111-F25V00 ISERA111-F25W00 ISERA111-F25X00 ISERA111-F25Y00 ISERA111-F25Z00 ISERA111-F29A00 ISERA111-F29B00 ISERA111-F29C00 ISERA111-F29D00 ISERA111-F29E00 ISERA111-F29F00 ISERA111-F29G00 ISERA111-F29H00 ISERA111-F29J00 ISERA111-F29K00 ISERA111-F29L00 ISERA111-F29M00 ISERA111-F29N00 ISERA111-F29P00 ISERA111-F29Q00 ISERA111-F29R00 ISERA111-F29S00 ISERA111-F29T00 ISERA111-F29U00 ISERA111-F29V00 ISERA111-F29W00 ISERA111-F29X00 ISERA111-F29Y00 ISERA111-F29Z00 ISERA113-F25A00 ISERA113-F25B00 ISERA113-F25D00 ISERA113-F25H00 ISERA113-F25J00 ISERA113-F25K00 ISERA113-F25L00 ISERA113-F29A00 ISERA113-F29B00 ISERA113-F29D00 ISERA113-F29H00 ISERA113-F29J00 ISERA113-F29K00 ISERA113-F29L00 ISERA112-F25A30 ISERA112-F25B30 ISERA112-F25C30 ISERA112-F25E30 ISERA112-F25F30 ISERA112-F25G30 ISERA112-F25A00 ISERA112-F25B00 ISERA112-F25C00 ISERA112-F25E00 ISERA112-F25F00 ISERA112-F25G00 ISERA112-F23A30 ISERA112-F23B30 ISERA112-F23C30 ISERA112-F23E30 ISERA112-F23F30 ISERA112-F23G30 ISERA112-F23A00 ISERA112-F23B00 ISERA112-F23C00 ISERA112-F23E00 ISERA112-F23F00 ISERA112-F23G00 ISERA112-F29A30 ISERA112-F29B30 ISERA112-F29C30 ISERA112-F29E30 ISERA112-F29F30 ISERA112-F29G30 ISERA112-F29A00 ISERA112-F29B00 ISERA112-F29C00 ISERA112-F29E00 ISERA112-F29F00 ISERA112-F29G00
ISAspher HPLC Columns, Phenyl
Kolony chromatografické
HPLC column of the ISAspher line guarantee the highest possible reliability in a wide range of applications.
  • The production of the silica used as well as the functionalisation are carried out according to highly developed and validated process
  • Reliable and proven
  • Flexibility and robustness for every HPLC application
This ensure unchanged properties of the stationary phases in terms of selectivity and consistent chemical properties.
Direct scale-up e.g. starting from a particle size of 5 µm to semi-preparative or preparative applications (up to 45 µMore Product Information