Tisk…
ISAspher HPLC Columns, Cyano
Dodavatel: ISERA
A111-223A00 A111-223B00 A111-223C00 A111-223D00 A111-223E00 A111-223F00 A111-223G00 A111-223H00 A111-223J00 A111-223K00 A111-223L00 A111-223M00 A111-223N00 A111-223P00 A111-223Q00 A111-223R00 A111-223S00 A111-223T00 A111-223U00 A111-223V00 A111-223W00 A111-223X00 A111-223Y00 A111-223Z00 A111-225A00 A111-225B00 A111-225C00 A111-225D00 A111-225E00 A111-225F00 A111-225G00 A111-225H00 A111-225J00 A111-225K00 A111-225L00 A111-225M00 A111-225N00 A111-225P00 A111-225Q00 A111-225R00 A111-225S00 A111-225T00 A111-225U00 A111-225V00 A111-225W00 A111-225X00 A111-225Y00 A111-225Z00 A111-229A00 A111-229B00 A111-229C00 A111-229D00 A111-229E00 A111-229F00 A111-229G00 A111-229H00 A111-229J00 A111-229K00 A111-229L00 A111-229M00 A111-229N00 A111-229P00 A111-229Q00 A111-229R00 A111-229S00 A111-229T00 A111-229U00 A111-229V00 A111-229W00 A111-229X00 A111-229Y00 A111-229Z00
ISERA111-223A00EA 8070 CZK
ISERA111-223A00 ISERA111-223B00 ISERA111-223C00 ISERA111-223D00 ISERA111-223E00 ISERA111-223F00 ISERA111-223G00 ISERA111-223H00 ISERA111-223J00 ISERA111-223K00 ISERA111-223L00 ISERA111-223M00 ISERA111-223N00 ISERA111-223P00 ISERA111-223Q00 ISERA111-223R00 ISERA111-223S00 ISERA111-223T00 ISERA111-223U00 ISERA111-223V00 ISERA111-223W00 ISERA111-223X00 ISERA111-223Y00 ISERA111-223Z00 ISERA111-225A00 ISERA111-225B00 ISERA111-225C00 ISERA111-225D00 ISERA111-225E00 ISERA111-225F00 ISERA111-225G00 ISERA111-225H00 ISERA111-225J00 ISERA111-225K00 ISERA111-225L00 ISERA111-225M00 ISERA111-225N00 ISERA111-225P00 ISERA111-225Q00 ISERA111-225R00 ISERA111-225S00 ISERA111-225T00 ISERA111-225U00 ISERA111-225V00 ISERA111-225W00 ISERA111-225X00 ISERA111-225Y00 ISERA111-225Z00 ISERA111-229A00 ISERA111-229B00 ISERA111-229C00 ISERA111-229D00 ISERA111-229E00 ISERA111-229F00 ISERA111-229G00 ISERA111-229H00 ISERA111-229J00 ISERA111-229K00 ISERA111-229L00 ISERA111-229M00 ISERA111-229N00 ISERA111-229P00 ISERA111-229Q00 ISERA111-229R00 ISERA111-229S00 ISERA111-229T00 ISERA111-229U00 ISERA111-229V00 ISERA111-229W00 ISERA111-229X00 ISERA111-229Y00 ISERA111-229Z00
ISAspher HPLC Columns, Cyano
Kolony chromatografické
HPLC column of the ISAspher line guarantee the highest possible reliability in a wide range of applications.
  • The production of the silica used as well as the functionalisation are carried out according to highly developed and validated process
  • Reliable and proven
  • Flexibility and robustness for every HPLC application
This ensure unchanged properties of the stationary phases in terms of selectivity and consistent chemical properties.
Direct scale-up e.g. starting from a particle size of 5 µm to semi-preparative or preparative applications (up to 45 µMore Product Information