Tisk…
Laboratory blenders, MiniMix® 100
Dodavatel: INTERSCIENCE
011230 013230
INSC011230EA 99270 CZK
INSC011230 INSC013230
Laboratory blenders, MiniMix® 100
Homogenizátory Mísiče laboratorní (blenders)
Laboratory blender for sample volumes of 5 to 80 ml.
  • Multifunction digital screen
  • Variable speed and time
  • Variable power, adjustable and removable paddles
  • Q-Tight® closing
  • MiniMix® 100 W CC® has a glass window in the door to observe the blending process