Tisk…
Anti-IgG Rabbit Polyclonal Antibody (HRP (Horseradish Peroxidase))
Dodavatel: ImmunoReagents
IR2530
IMMRIR2530EA 3590 CZK
IMMRIR2530
Anti-IgG Rabbit Polyclonal Antibody (HRP (Horseradish Peroxidase))
Protilátky
ImmunoReagents’ Horseradish Peroxidase (HRP) antibody conjugates are the most popular conjugates due to their high level of sensitivity, stability, and low costs. HRP conjugates are an ideal choice for use in ELISA, Immunohistochemistry (IHC), and Western blotting experiments.
  • Origin: US
ImmunoReagents' antibodies are suitable for most immunoassay formats and purified, cross adsorbed and conjugated based on the final product specfications.
Products will be accompanied with a standard product CoA to include application, purity and cross reactivityMore Product Information