Tisk…
Anti-IgG Goat Polyclonal Antibody (DyLight® 488)
Dodavatel: ImmunoReagents
IR2238
IMMRIR2238EA 3960 CZK
IMMRIR2238
Anti-IgG Goat Polyclonal Antibody (DyLight® 488)
Protilátky
The ImmunoReagents' DyLight® 488 conjugates provide exceptional performance for a wide array of fluorescence-base applications at a lower cost than other 488-range dyes. DyLight® 488 shows greater photostability than traditional fluorescein-based dyes and has a greater range of pH insensitivity (pH 4 to 9). DyLight® 488 conjugates are available for use in fluorescence microscopy, flow cytometry, Western blotting, ELISA, high-content screening and other array platforms. Excitation/Emission = 493 nm/518 nm Emission Colour = Green (Similar Dyes: Alexa Fluor 488, Cy2, FITC)
  • Origin: US