Tisk…
Anti-IgG F(c) Goat Polyclonal Antibody (HRP (Horseradish Peroxidase))
Dodavatel: ImmunoReagents
IR1724
IMMRIR1724EA 3650 CZK
IMMRIR1724
Anti-IgG F(c) Goat Polyclonal Antibody (HRP (Horseradish Peroxidase))
Protilátky
ImmunoReagents Horseradish Peroxidase (HRP) antibody conjugates are the most popular conjugates due to their high level of sensitivity, stability, and low costs. HRP conjugates are an ideal choice for use in ELISA, Immunohistochemistry (IHC), and Western blotting experiments.
  • Origin: US
ImmunoReagents antibodies are suitable for most immunoassay formats and purified, cross adsorbed and conjugated based on the final product specfications.
Products will be accompanied with a standard product CoA to include application, purity and cross reactivityMore Product Information
Objednat
Produktová specifikace
Symbol antigenu IgG F(c)
Název antigenu Immunoglobulin G
Environmentally Preferable
Druh protilátky Sekundární
Klonovalita Polyclonal
Spojení HRP (Horseradish Peroxidase)
Reaktivita Human
Hostitel Goat
Izotyp IgG
Western blot Yes
ELISA Yes
Imunochemie Yes
ImunoFluorescence Yes
Posun gelu Yes
Průtoková cytometrie Yes
Fragment protilátky F(c)
Epitop F(c)
Imunogen Human IgG, Fc fragment
Formát Purified IgG
Formulář Lyophilized
Čištění Affinity purified using solid phaseHuman IgG
Koncentrace 1,0 mg/ml (E 1% at 280 nm = 13,0)
Křížová adsorbce Yes
Skladovací pufr 10 mM Sodium Phosphate, 0,15 M Sodium Chloride, pH 7,2, 1 % (w/v) BSA, Protease/IgG free
Skladovací teplota 2…8 °C
Transportní teplota 2…8 °C