Tisk…
Anti-IgG Donkey Polyclonal Antibody (HRP (Horseradish Peroxidase))
Dodavatel: ImmunoReagents
IR1146
IMMRIR1146EA 6330 CZK
IMMRIR1146
Anti-IgG Donkey Polyclonal Antibody (HRP (Horseradish Peroxidase))
Protilátky
ImmunoReagents’ Horseradish Peroxidase (HRP) antibody conjugates are the most popular conjugates due to their high level of sensitivity, stability, and low costs. HRP conjugates are an ideal choice for use in ELISA, Immunohistochemistry (IHC), and Western blotting experiments.
  • Origin: US
ImmunoReagents' antibodies are suitable for most immunoassay formats and purified, cross adsorbed and conjugated based on the final product specfications.
Products will be accompanied with a standard product CoA to include application, purity and cross reactivityMore Product Information