Tisk…
Dry vacuum and standard duty diaphragm pumps, MP
Dodavatel: Welch by Gardner Denver
411721 411422 411722 411011 411421 411522 411543 411544
ILMV411721EA 48410 CZK
ILMV411721 ILMV411422 ILMV411722 ILMV411011 ILMV411421 ILMV411522 ILMV411543 ILMV411544
Dry vacuum and standard duty diaphragm pumps, MP
Čerpadla Pumpy vakuové membránové
These non-chemical-resistant diaphragm pumps characterized by a compact design, long lifespan and oil-free evacuation are designed for aqueous and standard duty applications. The diaphragm series are well known for smooth operation. They are available in one or multistage configuration with controlled (ecoflex) or uncontrolled pumping speed. Ecoflex diaphragm pumps are equipped with a state-of-the-art vacuum control which allows the pumping speed to be varied.
  • Different formats are available (1, 2, 4 head diaphragm pumps)
  • Low weight and compact design
  • Can
More Product Information