Tisk…
Laboratory reactor systems, LR-2.ST the versatile
Dodavatel: IKA
20013293
IKAA20013293EA 508890 CZK
IKAA20013293
Laboratory reactor systems, LR-2.ST the versatile
Reaktory laboratorní
Modular laboratory reactor for the optimisation and reproduction of various chemical reactions, mixing and homogenisation processes on a lab scale. The system is characterised by the agitator mounting, which allows a safe transfer of the higher motor torque.
  • Suitable for vacuum operation
  • Solvent- and temperature-resistant perfluoro elastomer (FFPM) seals meet the sample
  • Infinitely variable speed
  • Torque trend display for measuring changes in viscosity
  • Microprocessor-controlled speed regulation, enables steady speed, also under load
  • Removable WiCo (wireless
More Product Information