Tisk…
Handheld digital microscopes, Dino-Lite Medical, CapillaryScope
Dodavatel: Dino-Lite / IDCP B.V.
MEDL4N5 MEDL4N5PRO MEDL4NPRO
630-2614EA 13590 CZK
630-2614 IDCPMEDL4N5PRO IDCPMEDL4NPRO
Handheld digital microscopes, Dino-Lite Medical, CapillaryScope
Mikroskopy
These handheld microscopes are designed for the examination of the capillaries (tiny blood vessels) of the skin at the nailrim. The devices can show the capillaries in high magnification, as well as the decrease in capillaries, knot formation in capillaries and bleeding. With this information doctors can diagnose the condition of the blood vessels in the rest of the body.
  • Images can be captured as still photos or videos and stored on any digital medium
  • Magnifications of 200 and around 500× assure the acquisition of clear pictures
  • Powered through USB
More Product Information