Tisk…

Gentamicin sulphate 50 mg/ml, Reagent Grade

Dodavatel: MP Biomedicals

Synonyms: NMD Inhibitor II, Garamycin, Gentamicin sulphate salt, Gentiomycin C

0916762J8
ICNA0916762J8EA 17990 CZK
ICNA0916762J8
Gentamicin sulphate 50 mg/ml, Reagent Grade
Gentamicin sulphate
MDL Number: MFCD00270181
CAS číslo: 1405-41-0

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR