Tisk…
Mineral Mixes, AIN-93G, MP Biomedicals
Dodavatel: MP Biomedicals
0296040002 0296040010 0296040050
ICNA0296040002EA 2700 CZK
ICNA0296040002 ICNA0296040010 ICNA0296040050
Mineral Mixes, AIN-93G, MP Biomedicals
Potřeby veterinární Krmivo pro zvířata
This mineral mix, part of AIN-93G rodent diet, is used for growth, pregnancy and lactational phases of rodents.
For complete diet composition, please see the product datasheet.