Tisk…

Dimethylsulfoxid ≥99% cell culture reagent

Dodavatel: MP Biomedicals
Varování

Synonyms: Methanesulphinylmethane, Methyl sulphoxide, Dimethyl(oxido)sulphur, DMSO

0219605525 0219605580 0219605590 0219605591
ICNA0219605525EA 1150 CZK
ICNA0219605525 ICNA0219605580 ICNA0219605590 ICNA0219605591
Dimethylsulfoxid ≥99% cell culture reagent
Dimethylsulfoxid

Dimethyl Sulfoxide is ideal for use as a cryoprotectant. Also used in chemical reactions, in polymerase chain reactions (PCR) as a PCR cosolvent to help improve yields, especially in long PCR, and as a cryoprotectant vitrification agent for the preservation of cells, tissues and organs. DMSO is used in cell freezing media to protect cells from ice crystal-induced mechanical injury. It is used for frozen storage of primary, sub-cultured, and recombinant heteroploid and hybridoma cell lines, embryonic stem cells (ESC), and hematopoietic stem cells. DMSO is frequently used in the combinations with BSA or fetal bovine serum (FBS). Used to enhance dermal absorption of many chemicals. A solvent for many organic and inorganic compounds including fats, carbohydrates, dyes, resins, and polymers. Used in antifreeze or hydraulic fluids. A cryopreservative for cell cultures. Used in the oxidation of thiols and disulfides to sulfonic acids.

Dimethyl sulfoxide (DMSO) is a highly polar aprotic organic reagent that has exceptional solvent properties for organic and inorganic chemicals.

Physical Apppearance: Liquid
Viscosity: 1.1 centipoises (27 °C)
Vapor Density: 2.7 (vs air)
Vapor Pressure: 0.55 hPa at 20 °C, 0.42 mmHg (20 °C)
Refractive Index: n20/D 1.479(lit.)
Solubility: Soluble in water (1 mL DMSO + 1 mL H2O), methanol, acetone, ether, benzene, chloroform

Vzorec: (CH₃)₂SO
Bod varu: 189 °C (lit.)
Bod tání: 16…19 °C (lit.)
Hustota: 1,104 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00002089
CAS číslo: 67-68-5
EINECS: 200-664-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=3ec1573e80c11ddf9254283ef76abf5fb6539c7e&vid=157e2a24c93041cbbb69833d226195e333b07665&prid=web&clid=SAPDAM

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR